Adapters for TX-1000 Swinging Bucket Rotor
Adapters for TX-1000 Swinging Bucket Rotor
Actual product may vary
Thermo Scientific
Adapters for TX-1000 Swinging Bucket Rotor
An assortment of adapters for outstanding flexibility
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityFor Use With (Equipment)
750073024 x 500 mL500mL Corning Bottle
75003697100 x 9/10 mL9/10mL Blood Collection Tube
75003709164 x 4.5/6 mL4.5/6mL Blood Collection Tube
75003829150 to 300 mL Small Blood BagsSmall bag/cell culture bags 4 x 2 bags (< 350mL)
750073014 x 1000 mL1000mL Bio-Bottle
750042534 x 500 mL500mL Nalgene Bottle
750053924 x 250 mL250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
750073034 x 6 mL Standard or 4 x 2 mL Deep-WellMicroplate carriers
750073044 x 750 mL750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter
750073058 x 250 mL250mL Thermo Scientific Nalgene Bottle
7500730696 x 15 mL15mL Conical Tube
Catalog number 75007302
HSN 84219100
Price (SGD)
1,462.00
EA
Add to cart
Capacity:
4 x 500 mL
For Use With (Equipment):
500mL Corning Bottle
Price (SGD)
1,462.00
EA
Add to cart
An assortment of adapters for outstanding flexibility
Specifications
TypeRotor Adapter
Tube Type500 mL Corning Bottle
Capacity4 x 500 mL
Max. Tube Diameter98 mm
Max. Tube Length133 mm
For Use With (Equipment)500mL Corning Bottle
Unit SizeEA