Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Fisher Chemical
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, isopropanol, 2-hydroxypropane, isopropyl alcohol, alkolave, hartosol, dimethylcarbinol, avantine, sec-propyl alcohol, petrohol, 2-propanol, 2.5L, Report AU at 254nm, Report AU at 230nm, Report AU at 210nm, Report AU at 220nm | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
4 L
Packaging:
View all
Glass Bottle
Catalog number A461-212
HSN 29051220
Price (SGD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Propanol
pH7
Melting Point-89.5°C
AbsorbanceReport AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
CAS67-63-0
View more
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm