Polypropylene Sterilizing Tray Covers
Polypropylene Sterilizing Tray Covers
Actual product may vary
Bel-Art
Polypropylene Sterilizing Tray Covers
Corrosion-proof polypropylene plastic trays and covers (sold separately). Bel Art™ Polypropylene Sterilizing Trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
Have Questions?
Catalog NumberDimensions (L x W x H)
FIS22-08727012 x 8 x 1 in.
Catalog number FIS22-087270
Price (SGD)
613.00
Each
Add to cart
Dimensions (L x W x H):
12 x 8 x 1 in.
Price (SGD)
613.00
Each
Add to cart
  • Corrosion-proof polypropylene
  • Rounded corners for easy cleaning and smooth, tapered sides for stacking.
  • Covers are 3.2 mm (0.125 in.) thick with a 25.4 mm (1 in.) skirt that fits snugly over the rim of the tray
  • Steam autoclavable at 121°C (250°F)
Specifications
Capacity (Metric)5 L
Dimensions (L x W x H)12 x 8 x 1 in.
ColorWhite
Height (Metric)2.54 cm
Length (Metric)30.5 cm
MaterialPolypropylene
TypeTray Cover
Width (Metric)20.3 cm
For Use WithCover fits 5L Sterilizing Tray, Mfr. No. H162600000
Length (English)12 in.
Height (English)1 in.
ShapeRectangular
Thickness3.2 mm
Width (English)8 in.