Vikem Vinyl Aprons
Actual product may vary
Vikem Vinyl Aprons
Vikem Vinyl Aprons
Bel-Art
Vikem Vinyl Aprons
Provides protection to personnel in hospitals, schools, laboratories and industrial plants.
Have Questions?
Catalog NumberLength (Metric)
FIS22-26131491 cm
Catalog number FIS22-261314
Price (SGD)
123.00
Each
Length (Metric):
91 cm
Price (SGD)
123.00
Each
  • Lightweight and comfortable
  • Made from 4mil thick vinyl
  • Stitched cotton binding securing all edges
  • Strong tie backs double reinforced with metal grommets through three layers of material at waist
  • Non-absorbent and chemically resistant
Specifications
TypeApron
SizeUniversal
MaterialVinyl
ColorClear
DisposableReusable
Closure TypeTies
SterilityNon-sterile
ConstructionStitched cotton binding securing all edges
Dimensions (L x W)91 x 69cm (36 x 27 in.)
Length (Metric)91 cm
Seam TypeSewn
Unit SizeEach