Stainless-Steel Spoons
Actual product may vary
Bel-Art
Stainless-Steel Spoons
Double-ended spoons offer two bowl sizes for convenient sample handling
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)
FIS22-2615471.6 mL, 3 mL21 cm
FIS14-429E2 mL, 8 mL30 cm
Catalog number FIS22-261547
Price (SGD)
277.00
Pack
Capacity (Metric):
1.6 mL, 3 mL
Length (Metric):
21 cm
Price (SGD)
277.00
Pack
Double-ended spoons offer two bowl sizes for convenient sample handling
  • Polished 304 stainless-steel construction
  • Solid, rounded shaft for easy handling
Specifications
AutoclavableAutoclavable
Capacity (Metric)1.6 mL, 3 mL
ColorSilver
DisposableReusable
Product TypeSpoon
Length (English)8.26 in.
IncludesDouble end with 1.6mL small spoon at one end and 3mL large spoon at other
Length (English) Handle5.8 in.
Length (Metric) Handle14.73 cm
MaterialStainless Steel
Length (Metric)21 cm
Unit SizePack