Contact Plate, Petri Dish Rack
Actual product may vary
Bel-Art
Contact Plate, Petri Dish Rack
Safely transports, incubates and stores up to 60 contact plates with lids (up to 72 mm diameter) or 42 Petri dishes with lids (up to 10 mm diameter) to streamline benchtop sample processing.
Have Questions?
Catalog NumberPositionsHolds
FISS9013842Petri Dishes
Catalog number FISS90138
Price (SGD)
-
Positions:
42
Holds:
Petri Dishes
  • Compatible with RODAC™ and BD™ contact plates or 100 mm Petri dishes and lids
  • Rack is configured in 6 columns with bottom-access slots for easy, safe removal of items
  • Center divider has a built-in handle with ample space for labeling
  • Up to four racks can be stacked together and interlocked to prevent accidental knock over
  • A small, stable base preserves bench top space
  • Made from white polypropylene that is steam autoclavable at 121°C (250°F)
  • Racks are shipped flat and are easily assembled
Specifications
AutoclavableAutoclavable
ColorWhite
HoldsPetri Dishes
MaterialPolypropylene
Positions42
ShapeRectangular
StackableYes
SterilityNon-sterile
For Use With (Equipment)Petri Dishes w/ Lids (100 mm)
Dimensions (L x W x H)13 x 8.25 x 7 in.
Length (Metric)33 cm
Length (English)13 in.
Width (English)8.25 in.
Height (English)7 in.
IncludesCenter divider with handle
Product TypePetri Dish Rack
Diameter (Metric) Inner100 mm
Height (Metric)17.8 cm
Array2 x 3
Width (Metric)21 cm
Unit SizeEach