LO-tox™ Lab Thermometer
Actual product may vary
Fisherbrand
LO-tox™ Lab Thermometer
LO-tox™ lab thermometer without calibration certificate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)SubdivisionImmersion TypeLength (Metric)
FKS13068593-10°C to +330°C2°CPartial Immersion305 mm
FKS13008593-10°C to +50°C0.5°CPartial Immersion305 mm
FKS13018593-20°C to +110°C1°CPartial Immersion305 mm
FKS13028593-20°C to +150°C1°CPartial Immersion305 mm
FKS11907341-10°C to +260°C1°CPartial Immersion405 mm
FKS13008603-1°C to +51°C0.1°CTotal Immersion405 mm
FKS13058603-10°C to +101°C0.2°CPartial Immersion405 mm
Catalog number FKS13068593
Price (SGD)
27.00
Each
Temperature Range (Metric):
-10°C to +330°C
Subdivision:
2°C
Immersion Type:
Partial Immersion
Length (Metric):
305 mm
Price (SGD)
27.00
Each
Read temperatures quickly and accurately with blue spirit against a white background. Brannan™ Lo-Tox™ Spirit-filled partial Immersion Thermometer is manufactured to traceable standards and includes a blue non-roll top and safety suspension ring.
  • General purpose/stirring thermometer with a low toxic, environmentally friendly filling
  • Easy to read against background
  • Each thermometer is manufactured to traceable standards
Specifications
For Use With (Application)General use
Immersion TypePartial Immersion
IncludesBrannan non roll end cap
Length (Metric)305 mm
MaterialGlass
Thermometer FillBrown Spirit
Product TypeLaboratory Thermometer
Max. Temperature (English)626°F
Max. Temperature (Metric)330°C
Min. Temperature (English)14°F
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +330°C
For Use With (Equipment)General Use
Accuracy±2 at 90°C
Subdivision2°C
Immersion Length76 mm
Unit SizeEach