Microgard™ 2000 Standard Lab Coats with Collar
Actual product may vary
Fisherbrand
Microgard™ 2000 Standard Lab Coats with Collar
Constructed of microporous polyethylene laminate. Ansell Microgard™ 2000 Standard Lab Coats with Collar are approved for partial body chemical protection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS15586534X-Large
FKS15566534Medium
FKS15576534Large
Catalog number FKS15586534
Price (SGD)
-
Size:
X-Large
  • Front zip fastening
  • Left breast pocket
  • Lower right pocket
  • Bound seams
  • Excellent liquid penetration resistance and barrier to fine particulates (>0.01 microns*)
  • Silicone-free for paint spraying applications
  • Low linting to reduce the risk of fibre contamination in some critical areas
  • Anti-static according to EN 1149-5
Specifications
Product TypeGeneral Purpose Lab Coat
SizeX-Large
GenderUnisex
MaterialMicroporous Polyethylene Laminate (Non-woven)
ColorWhite
Closure TypeZipper
Cuff StyleElastic
DisposableLimited-use
SterilityNon-sterile
ResistanceChemicals, Infectious Agents
StyleMid-length
No. of Pockets2
Latex FreeYes
Seam TypeBound
Certifications/Compliance (Europe)Cat III Partial Body Protection Type PB (6)-B, EN14126, EN1149-1
Unit SizePack