Ethylenediaminetetraacetic Acid, Di Na Salt Dihydr. (Crystalline Powd./Electrophor.), Fisher BioReagents™
For minimization of metal ion impurities in reaction buffers
Have Questions?
Change view
Quantity:
1 kg
500 g
Packaging:
View all
Poly Bottle
Catalog number GEEBP120-1
Price (SGD)
209.00
Each
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
209.00
Each
Specifications
Chemical Name or MaterialEthylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
CAS6381-92-6
ChemAlert Storage SymbolGray
Health Hazard 2CAUTION!
Decomposition Information>252°C
View more
For minimization of metal ion impurities in reaction buffers
The disodium salt of EDTA is a chelating agent for metal ions and is used extensively in molecular biology to minimize metal ion impurities in reaction buffers.