Akura™ 96 Spheroid Microplates
Akura™ 96 Spheroid Microplates
Actual product may vary
Akura™ 96 Spheroid Microplates
Akura™ 96 Spheroid Microplates
InSphero
Akura™ 96 Spheroid Microplates
Automation compatible, secure handling, and easy observation. The Akura™ 96 Spheroid Microplate is designed for lossless spheroid/organoid formation and handling as well as reliable culturing and analysis.
Have Questions?
Catalog NumberQuantity
ISHCS-PB135 Pk.
Catalog number ISHCS-PB13
Price (SGD)
497.00
Special offer
Ends: 31-Mar-2026
859.00
Save 362.00 (42%)
PK
Add to cart
Quantity:
5 Pk.
Price (SGD)
497.00
Special offer
Ends: 31-Mar-2026
859.00
Save 362.00 (42%)
PK
Add to cart
The one-and-only 96-well spheroid plate with InSphero's unique features specially designed for lossless spheroid/organoid formation and handling as well as reliable culturing and analysis.
  • Unique SureXchange™ ledge engineered to prevent unintended spheroid/organoid aspiration.
  • Spheroid/organoid morphology and functionality is preserved in long-term culture by unique ULA coating.
  • Enhanced imaging speed and resolution through flat bottom well.
  • Improved spheroid/organoid formation with the Akura™ Tilting Stand by keeping plate at 30 degrees during cell aggregation.
  • Individually wrapped Akura™ 96 Spheroid Microplates with lid, made of cyclo olefin polymer, high-efficiency ULA coating.
Specifications
BarcodeNo Barcode
FormatSBS Standard
Array12 x 8
ColorClear
DisposableSingle-use
Product TypeULA Microplate
MaterialCyclic Olefin Copolymer
Volume (Metric) Well250 μL
No. of Wells96
Volume (Metric) Working70 μL
SterilitySterile
Surface TreatmentUltra-low Binding
Well ShapeRound, Flat Bottom (F-well)
Dnase FreeYes
Quantity5 Pk.
RNase FreeYes
Fit for PurposeSterile
Unit SizePK