Hand Soaps and Sanitizers
Actual product may vary
Micronova
Hand Soaps and Sanitizers
Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeSterility
MICIC-110Hand and Glove SoapNon-sterile
MICIC-210Hand SanitizerNon-sterile
MICIC-220Gloved Hand and Surface SanitizerSterile
Catalog number MICIC-110
Price (SGD)
-
Product Type:
Hand and Glove Soap
Sterility:
Non-sterile
Soaps and sanitizers formulated with highly effective, non-irritating ingredients are ideal for frequent use in clean and controlled environments. Designed for use with the MicroDispenser™. Include a saddle pump.
Hand care products are gentle and effective with built-in skin emollients for frequently washed hands. The 500 mL saddle pump bottles can be used as stand-alone dispensers or with the M-Zone™ MicroDispenser™ Touch-Free Soap Dispenser (ICDISP-3) to ensure uncontaminated results.
  • NovaClenz™ Instant Hand Sanitizer, made with 62% Ethanol to help prevent viral transfer. Effective against bacteria, including: pseudomonas, salmonella and staphylococcus. Effective in reducing nosocomial infections. Fortified with emollients to moisturize hands.
  • Hy-G-Clenz™ Antimicrobial Hand Soap, made with 0.13% Benzalkonium Chloride and natural emollients. Effective against a wide range of both gram-positive and gram-negative bacteria. Persistent activity up to three hours. pH balanced and contains natural moisturizers, ideal for frequent hand washing. Meets OSHA standards.
  • AquaHol™ Sterile Isopropyl Alcohol Solution, made with 70% Sterile IPA in 30% Wifi Quality Water, and meets USP requirements. Terminally filtered to 0.1 microns. Gamma Irradiated. Ideal for use over cleanroom gloves.
  • NovaDyne™ General Purpose Hand Soap, pH balanced gentle lotion hand soap. Made with built-in moisturizers/skin emollients, ideal for frequent hand washing.
  • LoNa™ Low Sodium Hand Soap, low ionic, low sudsing hand soap and glove cleanser, specially designed for the high-tech cleanroom; less than 100 ppb sodium. pH balanced and clean enough to be used on the face.
Specifications
Product TypeHand and Glove Soap
Color-codedNo
FormViscous Liquid
pH5.5 to 6.5
ScentFloral
SterilityNon-sterile
AppearanceClear to Straw
AutoclavableNot Autoclavable
Boiling Point100°C
Care InstructionsRub hands together for 20 seconds. Rinse thoroughly with warm running water.
Certifications/ComplianceCompliant with OSHA standards.
ColorPink
CompositionpH Balanced General Purpose Hand Soap
Container TypeHDPE
Density1.02 to 1.04
Dimensions (L x W x H)7.75 x 1.75 x 3.75
DisposableSingle-use
FlammableNo
For Use With (Application)Hand Soap
Height (English)7.75 in.
Height (Metric)19.69 cm
Length (English)3.75 in.
Length (Metric)9.53 cm
Packaging6 bottles per poly-lined case
Quantity6/Cs.
Resistant ToChemical
Size500 mL
SolubilityEasily soluble in cold water and hot water.
Volume (English)16.9 oz.
Volume (Metric)500 mL
Weight (English)16.9 oz.
Weight (Metric)0.5 kg
Width (English)1.75 in.
Width (Metric)4.45 cm
Unit SizeCase
Contents & Storage
Store at ambient temperature. Avoid direct sunlight. Empty containers retain product residue and can be hazardous. Do not reuse container.