Check Order Status
3-(4,5-Dimethyl-2-thazolyl)-2,5-diphenyl-2H-tetrazolium bromide, ∽98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
3-(4,5-Dimethyl-2-thazolyl)-2,5-diphenyl-2H-tetrazolium bromide, ∽98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210222701
1 g
Catalog number
MPB0210222701
Price (SGD)
228.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
228.00
EA
Add to cart
Specifications
Chemical Name or Material
3-(4, 5-Dimethyl-2-thazolyl)-2, 5-diphenyl-2H-tetrazolium bromide
CAS
298-93-1
Color
Orange-Yellow
Molecular Formula
C18H16BrN5S
MDL Number
MFCD00011964,MFCD00066662
View more