▸
Organic compounds
▸
Organic acids and derivatives
▸
Organic carbonic acids and derivatives
▸
Urea, Reagent grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Urea, Reagent grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB02103209.5
500 g
Catalog number
MPB02103209.5
Price (SGD)
69.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
69.00
Each
Add to cart
Specifications
Chemical Name or Material
Urea
CAS
57-13-6
Color
White
Density
1.335g/cm
3
at 20°C
Molecular Formula
CH4N2O
View more