Potassium Phosphate Monobasic, MP Biomedicals™
Potassium Phosphate Monobasic, MP Biomedicals™
Potassium Phosphate Monobasic, MP Biomedicals™
Potassium Phosphate Monobasic, MP Biomedicals™
MP Biomedicals
Potassium Phosphate Monobasic, MP Biomedicals™
Used as a component for a wide variety of media used in the culture of microorganisms. Potassium Phosphate Monobasic, MP Biomedicals is a reagent with high buffering capacity that helps maintain pH and supplies essential phosphate. It is a component in ph
Have Questions?
Catalog NumberQuantity
MPB02191430.5500 g
Catalog number MPB02191430.5
Price (SGD)
240.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
240.00
Each
Add to cart
Specifications
CAS7778-77-0
Recommended StorageStore at Room Temperature (15-30°C), desiccate.
Physical FormPowder
Quantity500 g
Molecular FormulaH2KO4P
View more
Potassium phosphate occurs in several forms: monobasic, dibasic, and tribasic (K3PO4). Most pH-neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.