Pen-Strep (10,000 IU/mL, 10 mg/mL), MP Biomedicals™
MP Biomedicals
Pen-Strep (10,000 IU/mL, 10 mg/mL), MP Biomedicals™
Pen-Strep (10,000 IU/mL, 10 mg/mL) is a double antibiotic solution containing 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.
Have Questions?
Catalog NumberQuantity
MPB091670249100 mL
Catalog number MPB091670249
Price (SGD)
66.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
66.00
Each
Specifications
Chemical Name or MaterialPenicillin-Streptomycin
TypePenicillin/Streptomycin
Quantity100 mL
Synonym200X Pen-Strep
Unit SizeEach
  • The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
  • This solution contains 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.