Blue Cardboard Cryobox
Blue Cardboard Cryobox
Actual product may vary
Tenak
Blue Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)
TENTE22082133 x 133 x 130 mm
TENTE22055133 x 133 x 75 mm
Catalog number TENTE22082
Price (SGD)
333.00
Pack
Add to cart
Dimensions (L x W x H):
133 x 133 x 130 mm
Price (SGD)
333.00
Pack
Add to cart
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
ColorBlue
MaterialCardboard
ShapeSquare
TypeCryo Box
Dimensions (L x W x H)133 x 133 x 130 mm
Unit SizePack