Check Order Status
Buffer Solution, pH 7.00 (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Buffer Solution, pH 7.00 (Certified), Fisher Chemical™
Potassium Phosphate Monobasic/Sodium Hydroxide
Have Questions?
Change view
Quantity:
500 mL
Packaging:
Poly Bottle
Catalog number
USFSB108500
Price (SGD)
41.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
41.00
EA
Add to cart
Chemical Identifiers
CAS
7778-77-0, 1310-73-2, 7732-18-5
Specifications
Chemical Name or Material
Buffer Solution, pH 7.00
CAS
7778-77-0
ChemAlert Storage Symbol
Gray
Melting Point
0°C
Color
Colorless
View more