Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Toluene, 99.85%, Extra Dry, AcroSeal™
Acros Organics
Toluene, 99.85%, Extra Dry, AcroSeal™
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 326981000
HSN 29023000
Price (SGD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.8 %
Water=<0.003 %