Tube Supports for Nessler Color Tubes
Tube Supports for Nessler Color Tubes
Actual product may vary
Bennett Wood Specialties
Tube Supports for Nessler Color Tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPositionsProduct TypeFor Use With (Equipment)
FIS07-065B12Tall Form Tube Rack100mL Nessler color tubes
FIS07-060A12Tall Form Tube Rack50mL Nessler color tubes
FIS07-065A6Tall Form Tube Rack50mL Nessler color tubes
Catalog number FIS07-065B
Price (SGD)
368.00
Each
Add to cart
Positions:
12
Product Type:
Tall Form Tube Rack
For Use With (Equipment):
100mL Nessler color tubes
Price (SGD)
368.00
Each
Add to cart
Specifications
Product TypeTall Form Tube Rack
AutoclavableNot Autoclavable
HoldsNessler Tubes (100 mL)
MaterialWood
Positions12
For Use With (Equipment)100mL Nessler color tubes
Unit SizeEach