PROSAT™ PS-911 Presaturated Wipes
PROSAT™ PS-911 Presaturated Wipes
Actual product may vary
Fisherbrand
PROSAT™ PS-911 Presaturated Wipes
PROSAT™ wipes are an effective, efficient and convenient method of cleaning process tools and other equipment. PROSAT wipes are also ideal for wiping articles prior to pass through.
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-06420522.9 x 27.9 cm
Catalog number FIS19-064205
Price (SGD)
34.00
Pack
Add to cart
Dimensions (L x W):
22.9 x 27.9 cm
Price (SGD)
34.00
Pack
Add to cart
  • Presaturated with IPA and DI water blends in flat, resealable pouches individually lot-numbered for traceability
  • These specially designed pouches preserve cleanliness and solvent saturation levels
  • Solvent blend meets SEMI specifications for metals and contaminants
  • ISO Class 5 compatible
  • 30 wipes/pouch, 50 pouches/case
Specifications
Product TypePre-saturated Wipe
MaterialPolypropylene
Certifications/ComplianceISO Class 5, SEMI for metals/contaminants
DisposableSingle-use
Dimensions (L x W)22.9 x 27.9 cm
For Use With (Application)Ideal for routine cleaning and wiping instruments and equipment
For Use With (Equipment)Process tools and other equipment
Packaging Quantity30 Wipes/Pouch, 50 Pouches/Cs.
Saturation Solution70% IPA/ 30% DI Water