▸
Pipette Products
▸
Pipette Product Accessories
▸
Replacement Bores for Drummond Micropipetters
Actual product may vary
Drummond
Replacement Bores for Drummond Micropipetters
Replacement parts
Have Questions?
Change view
Catalog Number
Type
FIS21-169A
Replacement Bore (10 μL)
FIS21-169B
Replacement Bore (20 μL)
2 Options
Catalog number
FIS21-169A
Price (SGD)
47.00
Pack
Add to cart
Type:
Replacement Bore (10 μL)
Price (SGD)
47.00
Pack
Add to cart
Replacement Parts
Specifications
Color
Orange
For Use With
Drummond MicroPipette (10 &muL), Digital Microdispenser
Material
Glass
Type
Replacement Bore (10 μL)
For Use With (Equipment)
Drummond Micropipetter, all microdispensers