Fixed-Volume Microdispensers
Actual product may vary
Drummond
Fixed-Volume Microdispensers
Overall volumetric accuracy of ±1% at full volume
Have Questions?
Catalog NumberVolume (Metric)
FIS21-176B20 μL
Catalog number FIS21-176B
Price (SGD)
969.00
Each
Volume (Metric):
20 μL
Price (SGD)
969.00
Each
  • Contain borosilicate glass bores with stainless steel plungers, except the 1000μL unit which has precision-molded plastic bores
  • All plungers with capacities 25μL and larger have PTFE tips
  • The 5μL unit (Cat. No. 21-170-10) will deliver 1 to 5μL
Specifications
Accuracy±1%
ColorBlack
Number of Channels1
TypeFixed Volume Microdispenser
Volume (Metric)20 μL
For Use With (Application)Precision molded Plastic bores
Includes2 Plungers, 2 O-rings, 1 collet, 1 coil spring, 1 spring guide
Unit SizeEach