PCR Micropipets
Actual product may vary
Drummond
PCR Micropipets
Disposable, positive displacement micropipets with stainless steel plungers
Have Questions?
Catalog NumberVolume (Metric)
FIS21-179B1 to 10 μL
Catalog number FIS21-179B
Price (SGD)
244.00
Pack
Volume (Metric):
1 to 10 μL
Price (SGD)
244.00
Pack
  • Measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
  • Decontaminate at temperature of 400°F or higher
Specifications
For Use With (Application)To measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
Includes100 disposable bores, 100 disposable plungers
Quantity100 Micropipets
Volume (Metric)1 to 10 μL
TypePCR Micropipet