Check Order Status
Borosilicate Glass Round Spinbar™ Magnetic Stir Bars
Actual product may vary
Bel-Art
Borosilicate Glass Round Spinbar™ Magnetic Stir Bars
Useful for high temperature applications up to 274°C (525°F)
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric)
Length (English)
Diameter (English)
FIS22-067579
38 mm
9.5 mm
1.5 in.
0.37 in.
Catalog number
FIS22-067579
Price (SGD)
65.00
Each
Add to cart
Length (Metric):
38 mm
Diameter (Metric):
9.5 mm
Length (English):
1.5 in.
Diameter (English):
0.37 in.
Price (SGD)
65.00
Each
Add to cart
Useful for high temperature applications up to 274°C (525°F)
Completely smooth surface offers zero absorption
Easy to clean
Meet US Pharmacopoeia specifications for Type I borosilicate glass
Manufactured in a registered ISO 9001:2008 facility
Autoclave safe
Specifications
Autoclavable
Autoclavable
Certifications/Compliance
USP Type I, ISO 9001:2008
Color
Clear
Diameter (English)
0.37 in.
Length (English)
1.5 in.
Coating
Glass
Diameter (Metric)
9.5 mm
Length (Metric)
38 mm
Material
Magnet
Quantity
1
Shape
Sphere
Type
Magnetic Stir Bar