Check Order Status
Flea Micro Spinbar™ Magnetic Stirring Bars
Actual product may vary
Bel-Art
Flea Micro Spinbar™ Magnetic Stirring Bars
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric)
Shape
Coating
FIS22-067678
5 mm
2 mm
Octagon
PTFE, FDA-grade, USP Class VI
Catalog number
FIS22-067678
Price (SGD)
16.00
Each
Add to cart
Length (Metric):
5 mm
Diameter (Metric):
2 mm
Shape:
Octagon
Coating:
PTFE, FDA-grade, USP Class VI
Price (SGD)
16.00
Each
Add to cart
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
Small size bar for stirring in vessels such as vials, micro test tubes and centrifuge tubes
Alnico magnet encapsulated in FDA Grade
USP Class VI PTFE low friction coating
Durable and inert
Autoclave safe
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
Specifications
Autoclavable
Autoclavable
Certifications/Compliance
FDA, USP Class VI, ISO 9001:2008
Color
White
Diameter (English)
0.07 in.
Length (English)
0.19 in.
Coating
PTFE, FDA-grade, USP Class VI
Diameter (Metric)
2 mm
Length (Metric)
5 mm
Material
Alnico Magnet
Quantity
50
Shape
Octagon
Type
Magnetic Stir Bar, Micro FLEA