KIMAX™ Test Tubes with Lip and Marking Spot
Actual product may vary
Kimble
KIMAX™ Test Tubes with Lip and Marking Spot
Durable Kimax borosilicate tubes.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FISS30608A10 mm75 mm4 mL
Catalog number FISS30608A
Price (SGD)
-
Diameter (Metric):
10 mm
Length (Metric):
75 mm
Capacity (Metric):
4 mL
Quality glass test tubes with lip and ceramic enamel marking spot.
Specifications
Certifications/ComplianceASTM E982, Type I
Design TypePlain
Capacity (Metric)4 mL
Length (Metric)75 mm
MaterialBorosilicate Glass
Label TypeMarking Spot
Diameter (Metric)10 mm
Unit SizePack