Check Order Status
Stackable Petri Dishes
Actual product may vary
Kord
Stackable Petri Dishes
For standard microbiology and general laboratory use.
Have Questions?
Catalog Number
Diameter (Metric)
Height (Metric)
Well Count
No. per Pack
FISS43566
100 mm
15 mm
4
25/Sleeve
Catalog number
FISS43566
Price (SGD)
-
Diameter (Metric):
100 mm
Height (Metric):
15 mm
Well Count:
4
No. per Pack:
25/Sleeve
Stackable lid for stability
Polystyrene construction
Clear top and bottom viewing surfaces
Specifications
Closure Type
Fully Stackable
Color
Clear
Disposable
Single-use
Material
Polystyrene
Shape
Round
Sterility
Sterile
Type
Stackable Petri Dish
Well Count
4
For Use With (Application)
Standard microbiology and general laboratory use
No. per Pack
25/Sleeve
Height (Metric)
15 mm
Diameter (Metric)
100 mm