Fast Output Teaching Still
Actual product may vary
Dynalon
Fast Output Teaching Still
Produces 4L of distilled water per hour and is easy to use.
Have Questions?
Catalog number FISS50923
Price (SGD)
-
This still lets you produce quality distillate while demonstrating the distillation process through a crystal-clear borosilicate glass boiler and a high-efficiency coil condenser. It is easy to use and safe to operate. A 3kW chromium-plated heater boils the water and the high-efficiency condenser provides distilled water output at 25° to 35°C (minimum water supply flowrate 1L/min.).
  • Simple to install
  • Strong metal stand for either bench or wall mounting
  • Condenser plumbing connections are via screwhead connectors so hose can be fitted or removed safely
  • Distilled water is produced at a rate of 4L/hr.
  • Quality of effluent complies with ASTM™ Type 4 and the European Pharmacopoeia for Purified Water
  • Built-in thermostat protects heater in the event of water supply failure, resets it automatically when the still has cooled
  • Unit can be cleaned without cumbersome dismantling; cleaning agents can be added to boiler through built-in funnel
  • Stopcock provided for draining
Specifications:
  • 220/240V, 50/60Hz
  • Dimensions: 20 × 6 × 18 in. (50 × 15 × 45cm)
  • Weight: 13.25 lb. (6kg)
  • Replaceable components are available for this item
Specifications
Capacity (English)135.2 oz.
Capacity (Metric)4 L
Certifications/ComplianceASTM type 4
Conductivity2
Dimensions (L x W x H)19.7 x 5.9 x 17.7 in. (50 x 15 x 45 cm)
Electrical Requirements220/240 V 50/60 Hz
For Use With (Application)Produces 4L/Hour distilled water
Frequency50/60 Hz
Heating Capacity (Metric)3
Height (English)17.7 in.
Height (Metric)45 cm
IncludesCrystal-clear Borosilicate Glass boiler and a high-efficiency coil condenser, strong Metal stand
Length (English)19.7 in.
Length (Metric)50 cm
MaterialMetal
Pressure Range3 psi
Temperature Range (English)77°F to 95°F
Temperature Range (Metric)25°C to 35°C
Voltage220 V
Weight (English)13.25 lb.
Weight (Metric)6 kg
Width (English)5.9 in.
Width (Metric)15 cm
Flow Rate4
TypeMerit Water Still
Unit SizeEach