Check Order Status
Hybridization Oven Bottles and Accessories
Actual product may vary
UVP
Hybridization Oven Bottles and Accessories
Have Questions?
Change view
Catalog Number
Product Type
Material
FISUVP07019401
Hybridization Bottle (15 cm L)
Borosilicate Glass
FISUVP98006703
Rocker Tray
Stainless Steel
2 Options
Catalog number
FISUVP07019401
Price (SGD)
382.00
EA
Add to cart
Product Type:
Hybridization Bottle (15 cm L)
Material:
Borosilicate Glass
Price (SGD)
382.00
EA
Add to cart
Specifications
Closure Material
Polypropylene, PTFE
Color
clear
For Use With (Application)
DNA, RNA, or protein identification
For Use With (Equipment)
UVP hybridization ovens
Length (Metric)
15 cm
Material
Borosilicate Glass
Product Type
Hybridization Bottle (15 cm L)
Volume (Metric)
142.52 mL