Check Order Status
Replacement UV Light Tubes
Actual product may vary
UVP
Replacement UV Light Tubes
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Have Questions?
Catalog Number
Wavelength
Wattage
FISUVP34006801
365 nm
20 W
Catalog number
FISUVP34006801
Price (SGD)
307.00
EA
Add to cart
Wavelength:
365 nm
Wattage:
20 W
Price (SGD)
307.00
EA
Add to cart
Specifications
Wattage
20 W
Wavelength
365 nm
For Use With (Equipment)
Model XX-20BLB UV bench lamp
Length (English)
23 in.
Length (Metric)
584.2 mm
Packaging Quantity
1 Ea.
Product Type
Replacement UV Light Tube