Check Order Status
Quickfit™ Right Angle Tubing Adapter
Actual product may vary
Quickfit
Quickfit™ Right Angle Tubing Adapter
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
Have Questions?
Catalog Number
Cone Size
FKS11342035
19/26
Catalog number
FKS11342035
Price (SGD)
105.00
EA
Add to cart
Cone Size:
19/26
Price (SGD)
105.00
EA
Add to cart
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
With right angle connection for attachment of rubber hose etc
Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
Specifications
Cone Size
19/26
For Use With (Equipment)
Flexible tubing, Rubber hose
Includes
Detachable Plastic screw thread connector
Material
Borosilicate Glass
Product Type
Right angle tubing adapter