Dimethyl Sulfoxide, Fisher BioReagents™
Dimethyl Sulfoxide, Fisher BioReagents™
Dimethyl Sulfoxide, Fisher BioReagents™
Dimethyl Sulfoxide, Fisher BioReagents™
Fisher Chemical
Dimethyl Sulfoxide, Fisher BioReagents™
Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 % | CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
Catalog number GEEBP231-100
Price (SGD)
79.00
Each
Add to cart
Quantity:
100 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (SGD)
79.00
Each
Add to cart
Specifications
Chemical Name or MaterialDimethyl Sulfoxide
Melting Point18.4°C
Absorbance≤0.30 Absorbance Units at 280nm
CAS67-68-5
ChemAlert Storage SymbolGray
View more
Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %
DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations.