▸
Solvents
▸
Isopropanols
▸
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
For nucleic acid precipitation | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change view
Quantity:
500 mL
1 L
2.5 L
Catalog number
GEEBP2618-500
Price (SGD)
59.00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
59.00
Each
Add to cart
Specifications
Chemical Name or Material
Isopropanol
pH
7
Melting Point
-89.5°C
Absorbance
0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
CAS
67-63-0
View more
For nucleic acid precipitation
Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids.