Hazard Bags
Hazard Bags
Actual product may vary
Micronova
Hazard Bags
Low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
Have Questions?
Catalog NumberColorCapacity (Metric)Sterility
MICBL2-32YEIRBBYellow114 LSterile
Catalog number MICBL2-32YEIRBB
Price (SGD)
389.00
Special offer
Ends: 31-Mar-2026
477.00
Save 88.00 (18%)
CS
Add to cart
Color:
Yellow
Capacity (Metric):
114 L
Sterility:
Sterile
Price (SGD)
389.00
Special offer
Ends: 31-Mar-2026
477.00
Save 88.00 (18%)
CS
Add to cart
Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste
  • Low density polyethylene Hazard bags
  • Red or Yellow Biohazard and Hazardous Material bags
  • Biohazard bags are clearly marked with a Biohazard symbol in black ink
  • Hazardous material bags are labeled for waste segregation in the US
  • The 32YE liners work with the BLSSH-1515 BinLiner Stand
  • The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
Specifications
Capacity (Metric)114 L
Certifications/ComplianceCertificate of Conformance and Certificate of Irradiation Processing; ISO 9001 Certified Facility Meets ASTM D170910-6 Sterility Assurance Level In-Compliance with AAMI Guidelines for Sterilization.
DescriptionFive folded liners per double bag
Diameter (Metric) Outer51.8 cm
Diameter (English) Outer20.4 in.
Capacity (English)30 gal.
ColorYellow
DisposableSingle-use
Height (Metric)91.4 cm
Length (Metric)81.3 cm
Quantity50/Cs.
ShapeSTAR: Durable pleated bottom to fit round containers.
SterilitySterile
AutoclavableNot Autoclavable
For Use With (Application)Transporting Biomedical Waste (Non-Sharps)
For Use With (Equipment)BLSSH series BinStand or similar container
Thickness0.050 mm
Height (English)36 in.
MaterialLDPE
Product TypeBiohazard Waste Bag
Temperature (Metric)15°C to 30°C
Unit SizeCS