MicroMop™ Adapters
MicroMop™ Adapters
Actual product may vary
Micronova
MicroMop™ Adapters
These lightweight and durable, plastic string mop adapters work with a range of string mops and are available with the choice between a stainless steel handle attachment or an included plastic handle.
Have Questions?
Catalog NumberMaterial
MICMH-1Fiberglass, Polypropylene
Catalog number MICMH-1
Price (SGD)
104.00
Each
Add to cart
Material:
Fiberglass, Polypropylene
Price (SGD)
104.00
Each
Add to cart
The Micronova MicroMop™ String Mop Adapters work with a variety of string mops, large and small, and are available with a choice of two handle options. The QDMH-1 adapter, with a Quick Disconnect handle attachment, connects to the stainless steel SSU series handles. The MH-1 adapter includes an attached, 60-inch plastic handle.
  • Orange polypropylene string mop holders
  • Lightweight and durable
  • Holds a range of string mops
  • QDMH-1 Adapter features a Stainless Steel, Quick Disconnect Handle Attachment
  • MH-1 Adapter includes an attached 60-inch plastic coated handle
  • Autoclavable
Specifications
Product TypeAttached Plastic Handle
ColorOrange
Length (English)60 in.
SterilityNon-sterile
Length (Metric)152 cm
AutoclavableAutoclavable
DisposableReusable
Care InstructionsStore at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Use With (Equipment)MicroMop™ and PolyGen™ Series Mops
For Use With (Application)For Cleaning Large Surface Areas Quickly and Efficiently
Color-codedNo
MaterialFiberglass, Polypropylene
Unit SizeEach