▸
Organic compounds
▸
Organoheterocyclic compounds
▸
Lactims
▸
Cycloheximide, from Microbial, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Cycloheximide, from Microbial, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210018305
5 g
Catalog number
MPB0210018305
Price (SGD)
344.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
344.00
Each
Add to cart
Specifications
Chemical Name or Material
Cycloheximide
Name Note
from
Microbial
; ∽800μg/mg
CAS
66-81-9
Color
White
EINECS Number
200-636-0
View more