Putrescine, ∽98%, MP Biomedicals™
Putrescine, ∽98%, MP Biomedicals™
Putrescine, ∽98%, MP Biomedicals™
Putrescine, ∽98%, MP Biomedicals™
MP Biomedicals
Putrescine, ∽98%, MP Biomedicals™
Putrescine is a GABA precursor in many biological systems and synthon for amido-ureas
Have Questions?
Catalog NumberQuantity
MPB021004412525 g
Catalog number MPB0210044125
Price (SGD)
100.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
100.00
Each
Add to cart
Specifications
Chemical Name or MaterialPutrescine
CAS110-60-1
ColorColorless
Density0.877g/mL at 25°C
EINECS Number203-782-3
View more