Sodium Azide, ≥99%, (Crystalline), MP Biomedicals
Sodium Azide, ≥99%, (Crystalline), MP Biomedicals
Sodium Azide, ≥99%, (Crystalline), MP Biomedicals
Sodium Azide, ≥99%, (Crystalline), MP Biomedicals
MP Biomedicals
Sodium Azide, ≥99%, (Crystalline), MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB0210289180100 g
Catalog number MPB0210289180
Price (SGD)
114.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
114.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium Azide
CAS26628-22-8
Storage Note 1Not for export
Recommended StorageRoom Temperature
Physical FormCrystalline
View more