▸
Recombinant Proteins
▸
Albumin, Bovine, Fatty Acid-Free
Actual product may vary
MP Biomedicals
Albumin, Bovine, Fatty Acid-Free
Prepared by Fraction V method
Have Questions?
Change view
Catalog Number
Quantity
MPB0215240180
100 g
MPB0215240110
10 g
2 Options
Catalog number
MPB0215240180
Price (SGD)
2,028.00
Each
Add to cart
Quantity:
100 g
Price (SGD)
2,028.00
Each
Add to cart
Low fatty acids
Purity: >98%
pH: 7
For best cell culture performance, see alternate Mfr. No. 180575
Peters, T., Albumin:an overview and bibliography, Miles Laboratory, Research Products Division:Elkhart, IN. (1980).
Specifications
For Use With (Application)
Immunohematology, Serology
Molecular Weight (g/mol)
66.338
Name
Bovine Albumin
pH Range
5.2 to 7
Quantity
100 g
Source
Bovine
Storage Requirements
2° to 8°C
Form
Powder