Check Order Status
Ammonium Acetate Anhydrous, ACS Reagent Grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Ammonium Acetate Anhydrous, ACS Reagent Grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219140491
1 kg
Catalog number
MPB0219140491
Price (SGD)
180.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
180.00
Each
Add to cart
Specifications
Chemical Name or Material
Ammonium Acetate
CAS
631-61-8
Storage Note 1
Store dessicated.
Recommended Storage
Store at room temperature.
Quantity
1 kg
View more