Check Order Status
DURAN™ GL 25 Laboratory Glass Bottles, Clear, With PP Screw Cap w/o Pouring Ring
Actual product may vary
Schott Duran
DURAN™ GL 25 Laboratory Glass Bottles, Clear, With PP Screw Cap w/o Pouring Ring
UV protection up to approx. 500 nm wavelength. Unchanged DURAN™ properties within the bottle, as coloration is only to the outer surface
Have Questions?
Catalog Number
Capacity (Metric)
SCH218011453
25 mL
Catalog number
SCH218011453
Price (SGD)
92.00
CS
Add to cart
Capacity (Metric):
25 mL
Price (SGD)
92.00
CS
Add to cart
With easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic
Select sizes available with retrace code
Specifications
Capacity (Metric)
25 mL
Color
Clear
Material
Glass
Thread Size (Metric)
GL 25
Height (Metric)
74 mm
Product Type
Laboratory Bottle
Diameter (Metric)
36 mm