Product Image
Schott Duran
DURAN™ GL 32 Laboratory Glass Bottles
DURAN™ GL 32 Laboratory Glass Bottles offer proven DURAN™ borosilicate glass quality. Available as bottle only or complete with blue PP screw cap (integral lip seal) and pouring ring for drip-free, clean, and safe working.
Have Questions?
Catalog NumberHeight (Metric)
SCH21801175391 mm
Catalog number SCH218011753
Price (SGD)
117.00
Case
Height (Metric):
91 mm
Price (SGD)
117.00
Case
  • DURAN™ borosilicate glass construction for chemical resistance and durability
  • GL 32 screw thread for compatibility with standard DURAN™ closures
  • Available as bottle only or as a complete set with blue PP screw cap (integral lip seal) and PP pouring ring
  • Pouring ring enables drip-free pouring for clean and safe working
  • Service temperature of screw cap and pouring ring: up to +140°C
  • Typical applications: storage, sample preparation, and transport
  • Select sizes available with retrace code
Specifications
Capacity (Metric)50 mL
ColorClear
MaterialBorosilicate Glass (3.3)
SterilityNon-sterile
Thread Size (Metric)GL 32
Closure IncludedNo
GraduatedYes
MouthThreaded
Height (Metric)91 mm
Closure SizeGL 32
Diameter (Metric)46 mm
AutoclavableAutoclavable
Capacity (English)2 oz.
ShapeRound
DisposableReusable
Product TypeLaboratory Bottle
Unit SizeCase