DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring
Actual product may vary
Schott Duran
DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring
DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring combine proven DURAN™ borosilicate glass quality with a blue PP screw cap (GL 45, integral lip seal) and PP pouring ring for drip-free, safe pouring. Ideal for storage, sample preparation and transport.
Have Questions?
Catalog NumberCapacity (Metric)
SCH2180163572000 mL
Catalog number SCH218016357
Price (SGD)
1,951.00
Case
Capacity (Metric):
2000 mL
Price (SGD)
1,951.00
Case
  • DURAN™ borosilicate glass bottle with GL 45 thread
  • Supplied complete with blue polypropylene (PP1) screw cap featuring integral lip seal
  • Polypropylene (PP1) pouring ring enables drip-free pouring and clean, safe working
  • Service temperature of screw cap and pouring ring: up to +140°C
  • Typical applications: storage, sample preparation and transport
  • Select sizes available with retrace code
Specifications
Capacity (Metric)2000 mL
ColorClear
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
MaterialBorosilicate Glass (Type I)
SterilityNon-sterile
Thread Size (Metric)GL 45
Closure IncludedYes
GraduatedYes
MouthThreaded
Height (Metric)265 mm
Closure SizeGL 45
Diameter (Metric)136 mm
AutoclavableAutoclavable
Capacity (English)68 oz.
ShapeSquare
DisposableReusable
Product TypeLaboratory Bottle
Unit SizeCase