30 mm Cryobox Dividers
Actual product may vary
Tenak
30 mm Cryobox Dividers
Dividers for use with 50 mm high cryoboxes.
Have Questions?
Catalog NumberHoldsTube Array
TENTE22113Vials with 24 mm O.D.5 x 5 cells
Catalog number TENTE22113
Price (SGD)
76.00
Each
Holds:
Vials with 24 mm O.D.
Tube Array:
5 x 5 cells
Price (SGD)
76.00
Each
  • For use in cardboard cryoboxes
  • Holds cryogenic vials
  • Grid style divider
Specifications
TypeGrid Divider
For Use With (Equipment)50 mm high cryoboxes
HoldsVials with 24 mm O.D.
Tube Array5 x 5 cells
Height (Metric)30 mm
Unit SizeEach