▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Chloride (Granular/USP/FCC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Chloride (Granular/USP/FCC), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
Packaging:
Poly Bottle
Catalog number
USFS640500
Price (SGD)
48.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
48.00
Each
Add to cart
Specifications
Boiling Point
1461°C
Chemical Name or Material
Sodium Chloride
Melting Point
801°C
CAS
7647-14-5
ChemAlert Storage Symbol
Gray
View more